

2-(2-Propynyloxy)tetrahydro-2H-pyran 98%
:6089-04-9
:
:C₈H₁₂O₂
:MFCD00006604
:140.18 g/mol
:63…65 °C (9 mmHg)
:0.997 g/cm³
:59 °C
:Refrigerator
:1993
:3,III
:GHS07
:GHS02
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4560 to 1.4580 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together