

Pyridine 99+% for analysis
:110-86-1
:
:C₅H₅N
:MFCD00011732
:115…116 °C (760 mmHg)
:–42 °C
:0.978 g/cm³
:17 °C
:1282
:3,II
:79.1 g/mol
:GHS07
:GHS02
:Ambient
- Appearance:Clear liquid
- Water:≤0.1 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Non-volatile matter:≤0.002 %
- Ammonia (NH3):≤0.002 %
Frequently Bought Together