DL-Isoserine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:565-71-9
:
Formula:C₃H₇NO₃
MDL Number:MFCD00008138
:105.09 g/mol
Melting Pt:235 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.5 %
Specification Test Results
Frequently Bought Together