Order Entry
Czech Republic
ContactUsLinkComponent
Dimethyl-trans-1,2-cyclopropanedicarboxylate 99+%
Irritant
Dimethyl-trans-1,2-cyclopropanedicarboxylate 99+%
Catalog # ACRO255350050
CAS Number:  
undefined
Irritant
Dimethyl-trans-1,2-cyclopropanedicarboxylate 99+%
Catalog # ACRO255350050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4425 to 1.4445 (20°C, 589 nm)
  • GC:
    ≥99.0 %

Specification Test Results

About this item

 :
 :158.153 g/mol
Boiling Pt:101 °C (18 mmHg)
Density:1.12 g/cm³
 :GHS07
Storage Temperature:Ambient
CAS Number:826-35-7
Formula:C₇H₁₀O₄
MDL Number:MFCD00062806