Palatinose hydrate 98%
:343336-76-5
:MFCD11100373
:C₁₂H₂₂O₁₁·xH₂O
:MFCD00076094
:122…124 °C
:342.30 g/mol (anhy)
:Ambient
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Water:≤6 %
- HPLC:≥97.5 % (ELSD)
- Specific optical rotation:+97° to +101° (20°C, 589 nm) (c=1, water)
Frequently Bought Together