Order Entry
Czech Republic
ContactUsLinkComponent
2-Pentanone 99+%, AcroSeal®, purified by redistillation
Irritant
Flammable
2-Pentanone 99+%, AcroSeal®, purified by redistillation
Certificates
undefined
Irritant
Flammable
2-Pentanone 99+%, AcroSeal®, purified by redistillation

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3890 to 1.3910 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA

Specification Test Results

Frequently Bought Together