
N-Acetylethylenediamine 90%
Catalog # ACRO223170050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline solid
- Infrared spectrum:Conforms
- Melting point:45 to 50 °C
- GC:≥88 %
Specification Test Results
About this item
:
:102.14 g/mol
Boiling Pt:143 °C (10 mmHg)
Melting Pt:50 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05
CAS Number:1001-53-2
Formula:C₄H₁₀N₂O
MDL Number:MFCD00008163