Order Entry
Czech Republic
ContactUsLinkComponent
Sodium acetate, anhydrous 99% for biochemistry
Sodium acetate, anhydrous 99% for biochemistry
Certificates
undefined
Sodium acetate, anhydrous 99% for biochemistry

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 %
  • UV:
    (0.1 M in water)
  • Titration with HClO4:
    ≥98.5 %
  • Heavy metals (as Pb):
    ≤0.001 %

Specification Test Results

Related Information

Frequently Bought Together