Potassium dihydrogen phosphate 99+% for biochemistry
:7778-77-0
:
:KH₂PO₄
:MFCD00011401
:136.09 g/mol
:~ 100 °C (1013 hPa)
:253 °C
:2,3 g/cm³ (20 °C)
:Ambient
- Appearance:Colorless to white Crystalline powder or crystals
- Solubility:(10 % in water) Clear colorless
- UV:(0.1 M in water)
- Titration with NaOH:≥99.0 %
- Heavy metals (as Pb):≤0.001 %
Frequently Bought Together