Aluminium nitrate nonahydrate 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7784-27-2
:
Formula:Al(NO₃)₃·9H₂O
MDL Number:MFCD00149132
:375.13 g/mol
Melting Pt:73 °C
Density:1,72 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:5.1,III
:GHS05
Specification Test Results
- Appearance:White Crystalline powder or crystals and/or chunks
- Titration Complexometric:6.5 to 7.9 % (Al)
- Total trace metal impurities:≤10 ppm
Specification Test Results
Frequently Bought Together