Order Entry
Czech Republic
ContactUsLinkComponent
Manganese(II) acetate tetrahydrate 99.999% (trace metals basis)
Irritant
Manganese(II) acetate tetrahydrate 99.999% (trace metals basis)
Catalog # ACRO212560500
CAS Number:  
undefined
Irritant
Manganese(II) acetate tetrahydrate 99.999% (trace metals basis)
Catalog # ACRO212560500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Appearance:
    Pink Crystalline powder or crystals
  • Titration Complexometric:
    21.8 to 23.0 %
  • Total trace metal impurities:
    ≤10 ppm

Specification Test Results

About this item

 :
 :245.09 g/mol
Melting Pt:>300 °C
Density:1,58 g/cm³ (20 °C)
Flash Pt:>130 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:6156-78-1
Formula:(H₃CCOO)₂Mn·4H₂O
MDL Number:MFCD00062552