Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium metavanadate 99.996% (trace metals basis)
Toxic
Health
Environment
Ammonium metavanadate 99.996% (trace metals basis)
Certificates
undefined
Toxic
Health
Environment
Ammonium metavanadate 99.996% (trace metals basis)

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder
  • Total trace metal impurities:
    ≤40 ppm
  • Vanadium (V):
    42.4 to 44.6 %

Specification Test Results

Related Information

Frequently Bought Together