Order Entry
Czech Republic
ContactUsLinkComponent
 
Discandium trioxide 99.999% (trace metals basis)
Catalog # ACRO193960050
CAS Number:  
undefined
Discandium trioxide 99.999% (trace metals basis)
Catalog # ACRO193960050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Total trace metal impurities:
    ≤10 ppm

Specification Test Results

About this item

 :
 :137.91 g/mol
Melting Pt:>1000 °C
Density:3,86 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:12060-08-1
Formula:O₃Sc₂
MDL Number:MFCD00011223