
1,1'-Azobis(N,N-dimethylformamide) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10465-78-8
:
Formula:C₆H₁₂N₄O₂
MDL Number:MFCD00008318
:172.19 g/mol
Melting Pt:111…113.5 °C
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Melting point:104 to 114 °C
- GC:≥97.5 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together