
4-Phenyl-3-butyn-2-one 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1817-57-8
:
Formula:C₁₀H₈O
MDL Number:MFCD00008776
:144.17 g/mol
Boiling Pt:75…76 °C (1.1hPa)
Density:0.99 g/cm³
Flash Pt:95 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together