
2,3-Dimethyl-1,3-butadiene 98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:513-81-5
:
Formula:C₆H₁₀
MDL Number:MFCD00008595
:82.15 g/mol
Boiling Pt:68…69 °C (760 mmHg)
Melting Pt:–76 °C
Density:0.726 g/cm³
Flash Pt:–20 °C
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
UN:3295
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4370 to 1.4400 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:100 ppm BHT
Specification Test Results
Frequently Bought Together