

3-Methoxy-1-propanol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1589-49-7
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00039550
:90.12 g/mol
Boiling Pt:149…150 °C (760 mmHg)
Density:0.942 g/cm³
Flash Pt:38 °C
Storage Temperature:Ambient
UN:1987
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4120 to 1.4140 (20°C, 589 nm)
- GC:≥97.5 %
- Specific optical rotation:+8° to +11° (20°C, 589 nm) (c=1,CH3OH)
Specification Test Results
Frequently Bought Together