


Methanol 99.9%, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Boiling Pt:64.7 °C (760 mmHg)
Melting Pt:–98 °C
Density:0.791 g/cm³
Flash Pt:12 °C
UN:1230
ADR:3,II
:GHS06
:GHS08
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.05 %
- GC:≥99.85 %
- UV:at 207 nm A: ≤1.0
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Titrable acid:≤0.0003 meq/g
- Titrable base:≤0.0002 meq/g
Specification Test Results
Related Information
Frequently Bought Together