Order Entry
Czech Republic
ContactUsLinkComponent
 
Mineral oil (petroleum) for IR spectrophotometry, suitable for nujol mull preparations
    
 
undefined
Mineral oil (petroleum) for IR spectrophotometry, suitable for nujol mull preparations
    
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless Oily liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4620 to 1.4730 (20°C, 589 nm)
  • Density:
    (15°C) 830 to 865 kg/l
  • Viscosity:
    13 to 19 mm²/s (40°C)

 

 

Frequently Bought Together