
2-Aminoacetophenone hydrochloride 96%
Catalog # ACRO103100010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to yellow to beige to light brown Powder or crystals
- Infrared spectrum:Conforms
- Titration Argentometric:95.0 to 105.0 %
- HPLC:≥98.5 %
Specification Test Results
About this item
:
:171.63 g/mol
Melting Pt:194 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:5468-37-1
Formula:C₈H₁₀ClNO
MDL Number:MFCD00012873