ContactUsLinkComponent
VWR® Protein Precipitation Kit
VWR® Protein Precipitation Kit
Catalog # 1B1301-KIT
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Protein Precipitation Kit
Catalog # 1B1301-KIT
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Protein Precipitation Kit
Specifications
About this item
The Protein Precipitation Kit utilises a simple trichloroacetic acid (TCA) based method to precipitate proteins in complex mixtures. Only protein in the sample is pelleted, while interfering salts and surfactants remain in the supernatant. A gentle acetone wash removes TCA and co-precipitants allowing the end-user to recover the protein in a small volume of buffer.
  • Removal of interfering salts, detergents and buffers
  • Reproducible and consistent results
Ordering information: Kit contains: Protein precipitation kit Solution A; 25 ml, protein precipitation kit Solution B, 250 ml; and wash buffer, 125 ml.