ContactUsLinkComponent
Foetal bovine serum alternative, Alpha Calf Fraction, US Origin, HyClone™
Foetal bovine serum alternative, Alpha Calf Fraction, US Origin, HyClone™
Catalog # HYCLSH30212.03
undefined
Foetal bovine serum alternative, Alpha Calf Fraction, US Origin, HyClone™
Catalog # HYCLSH30212.03
Specifications
  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    Alpha Calf Fraction, US origin, without iron
  • Seq:
    06
  • Pk:
    500 ml
Specifications
About this item
HyClone™ Alpha Calf Fraction provides hybridoma users with a cost-effective medium supplement that promotes growth and antibody production.