Graduated pipettes, conformity certified, COLOR CODE
Supplier: TECHNOSKLO
Certificates
About this item
Sodium-potassium glass, class AS.
- Calibrated for discharge, whole volume
- Cannot be autoclaved at 121 °C
Certifications: According to ISO 648.
Frequently Bought Together