1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene 90%
:108468-00-4
:
:C₁₃H₂₀N₂O₂
:MFCD02683058
:236.3100 g/mol
:64…68 °C
:3263
:8,III
:GHS07
:GHS05
:Ambient
- Appearance:White to brown Crystalline powder or solid
- Infrared spectrum:Conforms
- GC:≥89.0 %
- NMR (1H-NMR):Consistent with structure
Frequently Bought Together