



Di-tert-butyl pyrocarbonate 1 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24424-99-5
:
Formula:C₁₀H₁₈O₅
MDL Number:MFCD00008805
:218.25 g/mol
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:1992
ADR:3,II
:GHS02
:GHS05
:GHS06
:GHS08
Specification Test Results
- Appearance:Clear colorless Solution
- Titration by coupling:0.90 to 1.10 M
Specification Test Results
Related Information
Frequently Bought Together