


2-Methyl-2-butene 2 M in THF, AcroSeal®
Catalog # 426391000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Strength:1.90 to 2.20 M
- Melting point:107 to 110 °C
- GC:≥98.5 %
Specification Test Results
About this item
:
:70.13 g/mol
Boiling Pt:39 °C (1013 hPa)
Melting Pt:–124 °C
Density:0.835 g/cm³
Flash Pt:–46 °C
UN:1993
ADR:3,II
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient
CAS Number:513-35-9
:26760-64-5
Formula:C₅H₁₀
MDL Number:MFCD00009276