
Methyl-2-amino-5-chlorobenzoate 95%
ACRO426160010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:Cream to beige to light brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:68 to 72 °C
- GC:≥94 %
- NMR (1H-NMR):Consistent with structure
:
:185.61 g/mol
:168…170 °C (22 mmHg)
:66…68 °C
:GHS07
:Ambient
:5202-89-1
:C₈H₈ClNO₂
:MFCD00007837