Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
Potassium-tert-butoxide 1 M in tert-butanol, AcroSeal®
 
undefined
 
 
 
Potassium-tert-butoxide 1 M in tert-butanol, AcroSeal®

 

  • Appearance:
    Colorless to amber Solution
  • Strength:
    0.95 to 1.10 M
  • Refractive index:
    1.5750 to 1.5780 (20°C, 589 nm)
  • GC:
    ≥97.5 %

 

Frequently Bought Together