



Tebbe Reagent 0.5 M in toluene, AcroSeal®
Catalog # 426061000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Dark red to purple Solution
- Strength:0.40 to 0.60 M
- Refractive index:1.4165 to 1.4195 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
CAS Number:67719-69-1
:
Formula:C₁₃H₁₈AlClTi
MDL Number:MFCD00151575
:284.62 g/mol
Density:0.927 g/cm³
Flash Pt:4 °C
UN:2924
ADR:3,II
:GHS07
:GHS08
:GHS05
:GHS02
Storage Temperature:Freezer