
4-Chloro-2-fluoro-5-nitrotoluene 98%
Catalog # ACRO426010050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Yellow to light brown Crystalline mass
- Infrared spectrum:Conforms
- Melting point:42 to 45 °C
- GC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:189.57 g/mol
Melting Pt:42…45 °C
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
CAS Number:18349-11-6
Formula:C₇H₅ClFNO₂
MDL Number:MFCD00134231