Order Entry
Czech Republic
ContactUsLinkComponent
Titanium(IV) chloride 0.09 M in hydrochloric acid 20%, AcroSeal®
Corrosive
Irritant
Health
Titanium(IV) chloride 0.09 M in hydrochloric acid 20%, AcroSeal®
Catalog # 428351000.
CAS Number:  
undefined
Corrosive
Irritant
Health
Titanium(IV) chloride 0.09 M in hydrochloric acid 20%, AcroSeal®
Catalog # 428351000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to light yellow Solution
  • Strength:
    0.08 to 0.10 M
  • Elemental Analysis:
    ≥98.5 % (AES)

Specification Test Results

About this item

 :
 :189.69 g/mol
Storage Temperature:Ambient
UN:3264
ADR:8,II
 :GHS05
 :GHS07
 :GHS08
CAS Number:7550-45-0
Formula:Cl₄Ti
MDL Number:MFCD00011267