ContactUsLinkComponent
Sulphur trioxide N,N-dimethylformamide complex 47+% (active SO₃)
Corrosive
Sulphur trioxide N,N-dimethylformamide complex 47+% (active SO₃)
Certificates
undefined
Corrosive
Sulphur trioxide N,N-dimethylformamide complex 47+% (active SO₃)
Specification Test Results
  • Appearance:
    Beige to light brown Crystals
  • Infrared spectrum:
    Conforms
  • Titration with NaOH:
    ≥47.0 % (as SO3)