Order Entry
Czech Republic
ContactUsLinkComponent
N-(4-Pentynyl)phthalimide 97%
Irritant
N-(4-Pentynyl)phthalimide 97%
Certificates
undefined
Irritant
N-(4-Pentynyl)phthalimide 97%

Specification Test Results

  • Appearance:
    White to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    87 to 91 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together