

Boron trifluoride dibutyl etherate (30 - 35% BF₃), AcroSeal®
Catalog # 428141000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to brown Liquid
- Infrared spectrum:Conforms
- Strength:30.0 to 35.0 % (Boron trifluoride)
Specification Test Results
About this item
:
Density:0.959 g/cm³
Flash Pt:70 °C
ADR:8,II
:198.04 g/mol
UN:2922
:GHS06
:GHS05
Storage Temperature:Refrigerator
CAS Number:593-04-4
Formula:BF₃·C₈H₁₈O
MDL Number:MFCD00013195