Order Entry
Czech Republic
ContactUsLinkComponent
DL-3-Phenyllactic acid 97%
DL-3-Phenyllactic acid 97%
Certificates
undefined
DL-3-Phenyllactic acid 97%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    94 to 98 °C
  • HPLC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information

Frequently Bought Together