Order Entry
Czech Republic
ContactUsLinkComponent
2-Methylbutane for HPLC
Irritant
Health
Environment
Flammable
2-Methylbutane for HPLC
Certificates
undefined
Irritant
Health
Environment
Flammable
2-Methylbutane for HPLC

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.02 % (Coulometric)
  • GC:
    ≥99.0 %
  • UV:
    1 cm cell vs HPLC-grade water
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0003 %

Specification Test Results

Related Information

Frequently Bought Together