

Magnesium ethanolate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2414-98-4
:
Formula:C₄H₁₀MgO₂
MDL Number:MFCD00009060
:114.43 g/mol
Melting Pt:270 °C (dec.)
UN:3205
ADR:4.2,II
:GHS07
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light grey Powder
- Infrared spectrum:Conforms
- Additional info:May contain up to 2.5% Mg(OH)2 and/or MgCO3
- Titration with HCl:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together