3-Bromobenzoyl chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1711-09-7
:
Formula:C₇H₄BrClO
MDL Number:MFCD00000669
:219.46 g/mol
Boiling Pt:74.5…75 °C (0.5 mmHg)
Density:1.666 g/cm³
Flash Pt:107 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5940 to 1.5960 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together