


tert-Butylmagnesium chloride 1.7 M in diethyl ether, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:677-22-5
:
Formula:C₄H₉ClMg
MDL Number:MFCD00000465
:116.87 g/mol
Boiling Pt:34 °C (760 mmHg)
Density:0.8 g/cm³
Flash Pt:–40 °C
UN:3399
ADR:4.3,I
:GHS07
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark brown Solution
- Titration Active base:24 to 26 %
Specification Test Results
Related Information
Frequently Bought Together