Organic Reference Standard, Caffeine (1 mg/ml) in Methanol
Catalog # ACRO393540010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:58-08-2
- Description:Caffeine (< 100 µg/ml) in methanol
- Formula:C₈H₁₀N₄O₂
- Description:Caffeine