




Zinc chloride 0.7 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7646-85-7
:
Formula:ZnCl₂
MDL Number:MFCD00011295
:136.3 g/mol
Boiling Pt:66 °C
Flash Pt:–22 °C
Storage Temperature:Refrigerator
UN:2924
ADR:3,II
:GHS05
:GHS09
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear to hazy colorless Solution
- Titration Argentometric:0.65 to 0.8 M
- Water:≤0.1 %
Specification Test Results
Related Information
Frequently Bought Together