



Tetrakis(hydroxymethyl)phosphonium chloride ∼75-85% in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:124-64-1
:
Formula:C₄H₁₂ClO₄P
MDL Number:MFCD00031687
Flash Pt:96 °C
Storage Temperature:Ambient
UN:2922
ADR:8,III
:190.56 g/mol
:GHS05
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to yellow-green Solution
- Infrared spectrum:Conforms
- Titration Argentometric:75 to 85 %
Specification Test Results
Frequently Bought Together