

Propionyl bromide 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:598-22-1
:
Formula:C₃H₅BrO
MDL Number:MFCD00000116
:136.98 g/mol
Boiling Pt:104 °C (760 mmHg)
Density:1.52 g/cm³
Flash Pt:52 °C
Storage Temperature:Ambient
UN:2920
ADR:8,II
:GHS05
:GHS02
Specification Test Results
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4525 to 1.4575 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together