
2-Bromo-6-nitrotoluene 98%
Katalogové číslo ACRO398420050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light beige to brown Low melting solid
- Infrared spectrum:Conforms
- Melting point:38 to 42 °C
- GC:≥97.5 %
Specification Test Results
About this item
:
:216.03 g/mol
Boiling Pt:143 °C (22 mmHg)
Melting Pt:38…41 °C
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
CAS Number:55289-35-5
Formula:C₇H₆BrNO₂
MDL Number:MFCD00009792