Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium-4-hydroxybenzoate
Sodium-4-hydroxybenzoate 98%
Sodium-4-hydroxybenzoate 98%
:
Thermo Fisher Scientific
Sodium-4-hydroxybenzoate 98%
:
Thermo Fisher Scientific
:
114-63-6
:
:
C₇H₆O₃
:
MFCD00016530
:
160.10 g/mol
:
300 °C
:
GHS07
:
Ambient
Appearance:
White to light beige Crystalline powder
Infrared spectrum:
Conforms
Titration with HClO4:
≥97.5 %
ACRO39152
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Sodium benzoate 99.0-100.5% Ph. Eur.
: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
: VWR Chemicals
Formaldehyde ≥37% in aqueous solution stabilised, GR ACS, Reag. Ph. Eur. analytical reagent, Sigma-Aldrich®
: Merck
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals