

2-(Methylthio)ethylamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18542-42-2
:
Formula:C₃H₉NS
MDL Number:MFCD00014825
:91.18 g/mol
Flash Pt:36 °C
UN:2734
ADR:8,II
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4950 to 1.4980 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together