3-Ethynylpyridine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 3-Pyridylacetylene
CAS Number:2510-23-8
:
Formula:C₇H₅N
MDL Number:MFCD02177459
Boiling Pt:83…84 °C (30 mmHg)
Molecular Weight:103.12 g/mol
Melting Pt:39…40 °C
Flash Pt:56 °C
Storage Temperature:Ambient
UN:1325
ADR:4.1 II
:GHS07
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to brown Low melting solid (May darken on storage)
- Infrared spectrum:Conforms
- GC:≥95 %
Specification Test Results
Frequently Bought Together