
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:564-10-3
:
Formula:C₄H₂F₆O₂
MDL Number:MFCD00464165
:196.048 g/mol
Melting Pt:50…53 °C
Storage Temperature:Ambient
ADR:8,III
:GHS07
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:49 to 53 °C
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together