
4-(Trifluoromethyl)benzenesulphonyl chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2991-42-6
:
Formula:C₇H₄ClF₃O₂S
MDL Number:MFCD00042422
:244.62 g/mol
Boiling Pt:118…120 °C (18 mmHg)
Melting Pt:30…34 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to light yellow Low melting solid
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together