
4-Iodo-1H-pyrazole 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3469-69-0
:
Formula:C₃H₃IN₂
MDL Number:MFCD00005244
:193.97 g/mol
Melting Pt:108…110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:107 to 112 °C
- HPLC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together